Anti-ART1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90271-T24-50 90271-T24-200 90271-T24-100
103637-974EA 106.62 USD
103637-974 103589-848 103589-846
Anti-ART1 Rabbit Polyclonal Antibody
Antibodies
Anti-ART1 Antibody

Type: Primary
Antigen: ART1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR