Anti-ART1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90271-RP01-400 90271-RP01-100 90271-RP01-200
103662-524EA 562.97 USD
103662-524 103662-522 103662-520
Anti-ART1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus ART1/CD296/ARTC1 (Catalog#90271-C08H; XP_005579037.1; Gln23-Asn294). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: ART1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR