Anti-CD86 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90270-RP01-400 90270-RP01-100 90270-RP01-200
103662-516EA 351.49 USD
103662-516 103662-514 103662-518
Anti-CD86 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus CD86 / B7-2 ( Catalog#90270-C08H; Q9BDM4; Met1-His239). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR