Anti-CD80 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90268-T16-50 90268-T16-200 90268-T16-100
103589-840EA 270.87 USD
103589-840 103589-842 103637-972
Anti-CD80 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus CD80/B7-1 (Catalog#90268-C08H; ; Met1-Asn242). CD80/B7-1 specific IgG was purified by Cynomolgus CD80/B7-1 affinity chromatography.

Type: Primary
Antigen: B7-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR