Anti-IL2RA Rabbit Polyclonal Antibody

Supplier: Sino Biological
90265-RP01-400 90265-RP01-200 90265-RP01-100
103662-092EA 270.2 USD
103662-092 103662-090 103662-094
Anti-IL2RA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus CD25 / IL2RA (Catalog#90265-C08H; H6WS54; Met1-Arg213). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR