Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TFRC Rabbit Polyclonal Antibody

Supplier: Sino Biological

90253-T16-200 90253-T16-100 90253-T16-50
103589-836EA 214.31 USD InStock
103589-836 103589-834 103637-970
Anti-TFRC Rabbit Polyclonal Antibody
Antibodies
Anti-Transferrin Receptor Anti

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty