Anti-ALCAM Rabbit Monoclonal Antibody [clone: 008]

Supplier: Sino Biological
90227-R008-100 90227-R008-50
103592-238EA 150.18 USD
103592-238 103662-068
Anti-ALCAM Rabbit Monoclonal Antibody [clone: 008]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD166 / ALCAM (Catalog#90227-C08H; G7NZQ8; Met1-Ala526).

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: 008
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR