Anti-PDGFRB Rabbit Polyclonal Antibody

Supplier: Sino Biological
90215-T24-50 90215-T24-100 90215-T24-200
103637-954EA 142.96 USD
103637-954 103592-222 103592-220
Anti-PDGFRB Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus PDGFRB / CD140b (Catalog#90215-C08H; XP_001107595.1; Met1-Phe530). PDGFRB / CD140b specific IgG was purified by Rhesus PDGFRB / CD140b affinity chromatography.

Type: Primary
Antigen: PDGFRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR