Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological

90209-T16-200 90209-T16-100 90209-T16-50
103589-822EA 214.31 USD InStock
103589-822 103589-820 103637-950
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Anti-Semaphorin 4D Antibody

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty