Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological

90209-RP01-200 90209-RP01-400 90209-RP01-100
103662-046EA 160.46 USD
103662-046 103662-050 103662-048
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Anti-Semaphorin 4D Antibody

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR