Anti-CD83 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90199-T16-200 90199-T16-100 90199-T16-50
103589-816EA 270.87 USD
103589-816 103589-818 103637-948
Anti-CD83 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus / Rhesus CD83 (Catalog#90199-C08H; F6WX37; Met1-Ala143). CD83 specific IgG was purified by Cynomolgus / Rhesus CD83 affinity chromatography.

Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR