Anti-IL6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90197-RP02-200 90197-RP02-50 90197-RP02-100
103589-814EA 142.96 USD
103589-814 103662-038 103662-036
Anti-IL6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus IL6/Interleukin-6 (Catalog#90197-CNAE; NP_001036198.2; Pro29-Met212). IL6/Interleukin-6 specific IgG was purified by Rhesus IL6/Interleukin-6 affinity chromatography.

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR