Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD53 Rabbit Monoclonal Antibody [clone: 104]

Supplier: Sino Biological

90189-R104-50 90189-R104-100
103589-806EA 128.15 USD InStock
103589-806 103662-020
Anti-CD53 Rabbit Monoclonal Antibody [clone: 104]
Antibodies
Anti-CD53 Antibody

Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: Clone: 104
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty