Anti-CD59 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90188-RP01-200 90188-RP01-100 90188-RP01-400
103662-510EA 160.46 USD
103662-510 103662-512 103662-018
Anti-CD59 Rabbit Polyclonal Antibody
Antibodies
Anti-CD59 Antibody

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR