Anti-MME Rabbit Polyclonal Antibody

Supplier: Sino Biological
90177-T60-200 90177-T60-50 90177-T60-100
103637-942EA 142.96 USD
103637-942 103589-798 103589-800
Anti-MME Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus CD10/Neprilysin/MME ( Catalog#90177-C07H; NP_001247743.1; Tyr52-Trp750). CD10/Neprilysin/MME specific IgG was purified by Rhesus CD10/Neprilysin/MME affinity chromatography.

Type: Primary
Antigen: CD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR