Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SELE Rabbit Monoclonal Antibody [clone: 105]

Supplier: Sino Biological

90169-R105-50 90169-R105-100
103589-784EA 128.15 USD InStock
103589-784 103662-490
Anti-SELE Rabbit Monoclonal Antibody [clone: 105]
Antibodies
Anti-E-Selectin Antibody

Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: Clone: 105
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty