Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-REG1B Rabbit Polyclonal Antibody

Supplier: Sino Biological

90163-RP01-200 90163-RP01-100 90163-RP01-400
103662-008EA 160.46 USD InStock
103662-008 103662-010 103662-012
Anti-REG1B Rabbit Polyclonal Antibody
Antibodies
Anti-REG1B Antibody

Type: Primary
Antigen: REG1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty