Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-REG1B Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological

90163-MM05-200 90163-MM05-50 90163-MM05-100
103662-006EA 214.31 USD InStock
103662-006 103589-776 103662-004
Anti-REG1B Mouse Monoclonal Antibody [clone: 05]
Antibodies
Anti-REG1B Antibody

Type: Primary
Antigen: REG1B
Clonality: Monoclonal
Clone: Clone: 05
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty