Anti-CCL21 Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological
90138-MM05-50 90138-MM05-200 90138-MM05-100
103589-752EA 142.96 USD
103589-752 103661-980 103661-982
Anti-CCL21 Mouse Monoclonal Antibody [clone: 05]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus CCL21/6Ckine (Catalog#90138-CNAB; NP_001028027.1; Met1-Pro131). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CCL21
Clonality: Monoclonal
Clone: 05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR