Anti-FGFR4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90134-RP01-100 90134-RP01-200 90134-RP01-400
103662-482EA 351.49 USD
103662-482 103661-976 103662-484
Anti-FGFR4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus FGFR4 / CD334 ( Catalog#90134-C08H; XP_001087243.1; Met1-Asp369). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: FGFR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR