Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL18RAP Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological

90122-MM05-200 90122-MM05-50 90122-MM05-100
103661-972 103589-736 103661-970
Anti-IL18RAP Mouse Monoclonal Antibody [clone: 05]
Anti-IL18RAP Antibody

Type: Primary
Antigen: IL18RAP
Clonality: Monoclonal
Clone: Clone: 05
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty