Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL18R1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90121-T48-50 90121-T48-100 90121-T48-200
103637-860EA 106.62 USD InStock
103637-860 103597-690 103596-704
Anti-IL18R1 Rabbit Polyclonal Antibody
Antibodies
Anti-IL18R1 Antibody

Type: Primary
Antigen: IL18R1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty