Anti-IFNA13 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90108-T48-50 90108-T48-100 90108-T48-200
103637-854EA 142.96 USD
103637-854 103597-678 103597-680
Anti-IFNA13 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus IFNA13 (Catalog#90108-C08H; G7NFW4; Met1-Glu190). IFNA13 specific IgG was purified by Cynomolgus IFNA13 affinity chromatography.

Type: Primary
Antigen: IFNA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR