Anti-TNFRSF1B Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
90102-R001-50 90102-R001-100
103661-954EA 271.4 USD
103661-954 103597-670
Anti-TNFRSF1B Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus TNFR2/CD120b/TNFRSF1B ( Catalog#XP_005544817.1/NP_001253134.1; F7EAE4; Met1-Asp257).

Type: Primary
Antigen: TNFR2
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR