Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFR2 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological

90102-MM01-100 90102-MM01-200 90102-MM01-50
103661-950EA 160.46 USD InStock
103661-950 103661-952 103597-668
Anti-TNFR2 Mouse Monoclonal Antibody [clone: 01]
Antibodies
Anti-TNFR2 Antibody

Type: Primary
Antigen: TNFR2
Clonality: Monoclonal
Clone: Clone: 01
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty