Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DcR2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90100-T24-200 90100-T24-100 90100-T24-50
103597-666EA 214.31 USD InStock
103597-666 103597-664 103637-850
Anti-DcR2 Rabbit Polyclonal Antibody
Antibodies
Anti-DcR2 Antibody

Type: Primary
Antigen: DcR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty