Anti-TNFRSF10D Rabbit Polyclonal Antibody

Supplier: Sino Biological
90100-RP01-400 90100-RP01-100 90100-RP01-200
103661-948EA 562.97 USD
103661-948 103661-944 103661-946
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus TRAIL R4 / CD264 / TNFRSF10D (Catalog#90100-C08H; XP_001107922.1; Met1-Leu206). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: DcR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR