Anti-IGFBP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90083-T60-200 90083-T60-100 90083-T60-50
103597-648EA 270.87 USD
103597-648 103597-650 103637-842
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus IGFBP-2/IGFBP2 (Catalog#90083-C08H; XP_001087071.1; Met1-Gln324). IGFBP-2/IGFBP2 specific IgG was purified by Rhesus IGFBP-2/IGFBP2 affinity chromatography.

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR