Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological

90083-MM06-50 90083-MM06-200 90083-MM06-100
103597-646EA 106.62 USD InStock
103597-646 103661-918 103661-916
Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 06]
Antibodies
Anti-IGFBP2 Antibody

Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: Clone: 06
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty