Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological
90083-MM05-50 90083-MM05-100 90083-MM05-200
103661-914EA 352.03 USD
103661-914 103597-644 103661-912
Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 05]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus IGFBP-2/IGFBP2 ( Catalog#90083-C08H; XP_001087071.1; Met1-Gln324). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR