Anti-NOV Rabbit Polyclonal Antibody

Supplier: Sino Biological
90081-RP01-100 90081-RP01-400 90081-RP01-200
103661-904EA 270.2 USD
103661-904 103661-906 103661-902
Anti-NOV Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus CCN3/NOV (IGFBP9) (Catalog#90081-C08B; NP_001253825.1; Met1-Met357). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR