Anti-XCL1 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological
90079-R001-100 90079-R001-50
103597-626EA 150.18 USD
103597-626 103662-474
Anti-XCL1 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus XCL1 ( Catalog#90079-C08B; ; Met1-Gly114).

Type: Primary
Antigen: Lymphotactin
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR