Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FGF1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90062-T16-50 90062-T16-100 90062-T16-200
103637-834EA 106.62 USD InStock
103637-834 103597-622 103597-624
Anti-FGF1 Rabbit Polyclonal Antibody
Antibodies
Anti-FGF1 Antibody

Type: Primary
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty