Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HER3 / ERBB3 Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological

90043-MM05-50 90043-MM05-200 90043-MM05-100
103597-612EA 106.62 USD InStock
103597-612 103662-460 103662-458
Anti-HER3 / ERBB3 Mouse Monoclonal Antibody [clone: 05]
Antibodies
Anti-HER3 / ERBB3 Antibody

Type: Primary
Antigen: HER3 / ERBB3
Clonality: Monoclonal
Clone: Clone: 05
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty