Anti-YWHAB Mouse Monoclonal Antibody [clone: 05]

Supplier: Sino Biological
90022-MM05-100 90022-MM05-200 90022-MM05-50
103661-896EA 352.03 USD
103661-896 103597-598 103661-894
Anti-YWHAB Mouse Monoclonal Antibody [clone: 05]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus 14-3-3 beta/YWHAB (Catalog#90022-CNCE; Q4R572-2; Met 2-Asn 244). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: 14-3-3 beta
Clonality: Monoclonal
Clone: 05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR