Anti-ERBB2 Mouse Monoclonal Antibody [clone: 10A7F7]

Supplier: Sino Biological
90020-MM01-100 90020-MM01-200 90020-MM01-50
103662-448EA 270.87 USD
103662-448 103597-594 103662-450
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 10A7F7]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus HER2/ErbB2 (Catalog#90020-K08H; XP_001090430.1; Met 1-Thr 652). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Her2 / ERBB2
Clonality: Monoclonal
Clone: 10A7F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR