Anti-CD32A Rabbit Monoclonal Antibody [clone: 085]

Supplier: Sino Biological

90016-R085-50 90016-R085-100
103597-588EA 128.15 USD
103597-588 103661-882
Anti-CD32A Rabbit Monoclonal Antibody [clone: 085]
Antibodies
Anti-CD32A Antibody

Type: Primary
Antigen: CD32A
Clonality: Monoclonal
Clone: Clone: 085
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR