Anti-FCGR2A Rabbit Monoclonal Antibody [clone: 043] PE (Phycoerythrin)

Supplier: Sino Biological
90016-R043-P-100 90016-R043-P-25
103632-468EA 222.2 USD
103632-468 103630-272
Anti-FCGR2A Rabbit Monoclonal Antibody [clone: 043] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus CD32a/FCGR2A/Fc gamma RIIA ( Catalog#90016-C08H; NP_001244229.1; Met1-Ile217) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD32A
Clonality: Monoclonal
Clone: Clone: 043
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR