Anti-CD32A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 04]

Supplier: Sino Biological

90016-MM04-A-25 90016-MM04-A-100
103629-552EA 107.69 USD
103629-552 103632-460
Anti-CD32A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 04]
Antibodies
Anti-CD32A Antibody (APC)

Type: Primary
Antigen: CD32A
Clonality: Monoclonal
Clone: Clone: 04
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR