Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD32A Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological

90016-MM02-200 90016-MM02-100 90016-MM02-50 90016-MM02-1
103661-870EA 214.31 USD InStock
103661-870 103661-868 103597-580 103637-820
Anti-CD32A Mouse Monoclonal Antibody [clone: 02]
Antibodies
Anti-CD32A Antibody

Type: Primary
Antigen: CD32A
Clonality: Monoclonal
Clone: Clone: 02
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty