Anti-FCGR2A Rabbit Polyclonal Antibody

Supplier: Sino Biological
90015-RP02-200 90015-RP02-50 90015-RP02-100
103661-862EA 352.03 USD
103661-862 103597-576 103661-860
Anti-FCGR2A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Cynomolgus CD32a / Fc gamma RIIA (rC CD32a / Fc gamma RIIA; Catalog#90015-C08H; AAL92096.1; Met 1-Ile 211). CD32a / Fc gamma RIIA specific IgG was purified by Cynomolgus CD32a / Fc gamma RIIA affinity chromatography.

Type: Primary
Antigen: Anti-CD32A Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR