Anti-FCGR2B Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological
90014-MM06-50 90014-MM06-200 90014-MM06-100
103597-568EA 142.96 USD
103597-568 103662-656 103662-654
Anti-FCGR2B Mouse Monoclonal Antibody [clone: 06]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus CD32b / FCGR2B / Fc gamma RIIB ( Catalog#90014-C08H; AAL92097.1; Met1-Pro217). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Anti-CD32B Antibody
Clonality: Monoclonal
Clone: 06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR