Anti-IL-18 Rabbit Polyclonal Antibody

Supplier: Sino Biological
90011-RP02-50 90011-RP02-100 90011-RP02-200
103661-838EA 160.18 USD
103661-838 103596-934 103661-840
Anti-IL-18 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-18 Antibody

Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR