Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL-18 Mouse Monoclonal Antibody [clone: 5G10E10]

Supplier: Sino Biological

90011-MM02-50 90011-MM02-100 90011-MM02-200
103596-930EA 106.62 USD InStock
103596-930 103661-832 103662-418
Anti-IL-18 Mouse Monoclonal Antibody [clone: 5G10E10]
Antibodies
Anti-IL-18 Antibody

Type: Primary
Antigen: IL-18
Clonality: Monoclonal
Clone: Clone: 5G10E10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Rhesus Monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty