Anti-IFNG Rabbit Polyclonal Antibody

Supplier: Sino Biological
90008-T16-50 90008-T16-200 90008-T16-100
103637-812EA 142.96 USD
103637-812 103597-562 103597-564
Anti-IFNG Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus IFN gamma (Catalog#90008-CNAE; NP_001028077.1; Gln24-Gln165). IFN gamma specific IgG was purified by Rhesus IFN gamma affinity chromatography.

Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR