Anti-IFNG Rabbit Polyclonal Antibody

Supplier: Sino Biological
90008-RP01-200 90008-RP01-400 90008-RP01-100
103661-822EA 229.88 USD
103661-822 103661-824 103661-820
Anti-IFNG Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rhesus IFN gamma (Catalog#90008-CNAE; NP_001028077.1; Gln24-Gln165). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR