Anti-TIM-1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90006-RP02-100 90006-RP02-200 90006-RP02-50
103661-816EA 160.46 USD
103661-816 103661-818 103597-556
Anti-TIM-1 Rabbit Polyclonal Antibody
Antibodies
Anti-TIM-1 Antibody

Type: Primary
Antigen: TIM-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR