Anti-HAVCR1 Mouse Monoclonal Antibody [clone: 5F8H3]

Supplier: Sino Biological
90006-MM01-50 90006-MM01-200 90006-MM01-100
103597-550EA 142.96 USD
103597-550 103662-412 103662-410
Anti-HAVCR1 Mouse Monoclonal Antibody [clone: 5F8H3]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus KIM-1 / TIM1 / HACVR1 (Catalog#90006-C07H; BAJ61041.1; Asp20-Val135). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TIM-1
Clonality: Monoclonal
Clone: 5F8H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR