Anti-CD155 Rabbit Polyclonal Antibody

Supplier: Sino Biological

90005-RP01-100 90005-RP01-400 90005-RP01-200
103661-800EA 106.62 USD
103661-800 103661-804 103661-802
Anti-CD155 Rabbit Polyclonal Antibody
Antibodies
Anti-CD155 / PVR Antibody

Type: Primary
Antigen: CD155
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR