Anti-SFTPD Rabbit Monoclonal Antibody [clone: 111]

Supplier: Sino Biological

90003-R111-50 90003-R111-100
103597-540EA 128.15 USD
103597-540 103661-774
Anti-SFTPD Rabbit Monoclonal Antibody [clone: 111]
Antibodies
Anti-Surfactant protein D Anti

Type: Primary
Antigen: SFTPD
Clonality: Monoclonal
Clone: Clone: 111
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rhesus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR